Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGGGGAAGTTCTGAATTAAGTC[A/G]TTGAGAGTGTTACCTGGGCATCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609042 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OPN5 - opsin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181744.3 | 167 | Intron | NP_859528.1 | |||
XM_017010409.1 | 167 | Intron | XP_016865898.1 | |||
XM_017010410.1 | 167 | Intron | XP_016865899.1 | |||
XM_017010411.1 | 167 | Intron | XP_016865900.1 | |||
XM_017010412.1 | 167 | UTR 5 | XP_016865901.1 | |||
XM_017010413.1 | 167 | Intron | XP_016865902.1 | |||
XM_017010414.1 | 167 | Intron | XP_016865903.1 | |||
XM_017010415.1 | 167 | UTR 5 | XP_016865904.1 | |||
XM_017010416.1 | 167 | Intron | XP_016865905.1 |