Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTCTGTTCCTTCCTCTCCTGC[A/C]CAGTGGTCAGGTAGTTGACGGCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137161 | ||||||||||||||||||||
Literature Links: |
GABRR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABRR1 - gamma-aminobutyric acid type A receptor rho1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256703.1 | 1516 | Missense Mutation | GGG,GTG | G,V 354 | NP_001243632.1 | |
NM_001256704.1 | 1516 | Missense Mutation | GGG,GTG | G,V 284 | NP_001243633.1 | |
NM_001267582.1 | 1516 | Missense Mutation | GGG,GTG | G,V 284 | NP_001254511.1 | |
NM_002042.4 | 1516 | Missense Mutation | GGG,GTG | G,V 371 | NP_002033.2 | |
XM_017010689.1 | 1516 | Missense Mutation | GGG,GTG | G,V 284 | XP_016866178.1 |