Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATCCCTGGGGGTAATCGCTTC[C/T]GTTTTTTCCAGCCCACTATTATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124450 MIM: 608276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDO - D-aspartate oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A16 - solute carrier family 22 member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033125.3 | 1266 | Silent Mutation | ACA,ACG | T,T 565 | NP_149116.2 | |
XM_005267184.3 | 1266 | Silent Mutation | ACA,ACG | T,T 395 | XP_005267241.1 | |
XM_011536204.2 | 1266 | Silent Mutation | ACA,ACG | T,T 522 | XP_011534506.1 | |
XM_011536205.2 | 1266 | UTR 3 | XP_011534507.1 | |||
XM_011536206.2 | 1266 | Silent Mutation | ACA,ACG | T,T 471 | XP_011534508.1 | |
XM_011536207.2 | 1266 | Silent Mutation | ACA,ACG | T,T 447 | XP_011534509.1 | |
XM_011536208.2 | 1266 | Silent Mutation | ACA,ACG | T,T 435 | XP_011534510.1 | |
XM_011536209.2 | 1266 | Silent Mutation | ACA,ACG | T,T 430 | XP_011534511.1 | |
XM_011536210.1 | 1266 | Intron | XP_011534512.1 | |||
XM_011536211.2 | 1266 | Silent Mutation | ACA,ACG | T,T 405 | XP_011534513.1 | |
XM_011536212.2 | 1266 | Silent Mutation | ACA,ACG | T,T 335 | XP_011534514.1 |