Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTTTTTGGTCCCTTCCTCCTG[G/T]AATGGAATCGTGGCGCTGCTGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CNKSR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNKSR3 - CNKSR family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173515.2 | 4545 | Missense Mutation | TTA,TTC | L,F 519 | NP_775786.2 | |
XM_011535485.2 | 4545 | Missense Mutation | TTA,TTC | L,F 509 | XP_011533787.1 |