Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACGGCGATATCTAGAATCCGGC[A/G]TAGTCCTGAGGAGAACAGGTACCGA
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600169 | |||||||||||||||||||||||||||||||||||
Literature Links: |
MICA PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
AMR
|
|||||
EUR - Not Available |
MICA - MHC class I polypeptide-related sequence A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000247.2 | 661 | Missense Mutation | ATA,GTA | I,V 199 | NP_000238.1 | |
NM_001177519.2 | 661 | Missense Mutation | ATA,GTA | I,V 199 | NP_001170990.1 | |
NM_001289152.1 | 661 | Missense Mutation | ATA,GTA | I,V 102 | NP_001276081.1 | |
NM_001289153.1 | 661 | Missense Mutation | ATA,GTA | I,V 102 | NP_001276082.1 | |
NM_001289154.1 | 661 | Missense Mutation | ATA,GTA | I,V 61 | NP_001276083.1 |