Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATACTTGATTTCCAGATGGTTG[A/C]GGTTTTTCTTTTTTAATATACTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS3ST5 - heparan sulfate-glucosamine 3-sulfotransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153612.3 | 3707 | UTR 3 | NP_705840.2 | |||
XM_006715379.2 | 3707 | UTR 3 | XP_006715442.1 | |||
XM_011535588.2 | 3707 | UTR 3 | XP_011533890.1 | |||
XM_017010470.1 | 3707 | UTR 3 | XP_016865959.1 | |||
XM_017010471.1 | 3707 | UTR 3 | XP_016865960.1 | |||
XM_017010472.1 | 3707 | UTR 3 | XP_016865961.1 | |||
XM_017010473.1 | 3707 | UTR 3 | XP_016865962.1 | |||
XM_017010474.1 | 3707 | UTR 3 | XP_016865963.1 |
LOC101927768 - uncharacterized LOC101927768 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |