Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTTCAGAATACCAGGAGCGATA[C/T]GGTTCTCGCCCCGTCTGGGCTGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615556 MIM: 611982 MIM: 603771 | ||||||||||||||||||||
Literature Links: |
ATAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAT1 - alpha tubulin acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS18B - mitochondrial ribosomal protein S18B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014046.3 | 361 | Silent Mutation | TAC,TAT | Y,Y 68 | NP_054765.1 |
PPP1R10 - protein phosphatase 1 regulatory subunit 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |