Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAGGAAGATGGCTATAAACAGA[A/G]TCCAGGAGGGTCTGGCTGGCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFRSF21 - TNF receptor superfamily member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014452.4 | 2371 | Silent Mutation | GAC,GAT | D,D 645 | NP_055267.1 | |
XM_017010744.1 | 2371 | Intron | XP_016866233.1 |