Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAATGCCACCAAAGAGTTAGCC[A/G]ATAAAAATGGAACTTACTGAACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL32 - kelch like family member 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286250.1 | 216 | Intron | NP_001273179.1 | |||
NM_001286251.1 | 216 | Intron | NP_001273180.1 | |||
NM_001286252.1 | 216 | Intron | NP_001273181.1 | |||
NM_001286254.2 | 216 | Intron | NP_001273183.1 | |||
NM_001323252.1 | 216 | Intron | NP_001310181.1 | |||
NM_001323253.1 | 216 | Intron | NP_001310182.1 | |||
NM_001323254.1 | 216 | Intron | NP_001310183.1 | |||
NM_001323255.1 | 216 | Intron | NP_001310184.1 | |||
NM_001323256.1 | 216 | Intron | NP_001310185.1 | |||
NM_001323257.1 | 216 | Intron | NP_001310186.1 | |||
NM_001323258.1 | 216 | UTR 5 | NP_001310187.1 | |||
NM_001323259.1 | 216 | Intron | NP_001310188.1 | |||
NM_001323260.1 | 216 | Intron | NP_001310189.1 | |||
NM_001323261.1 | 216 | Intron | NP_001310190.1 | |||
NM_001323262.1 | 216 | Intron | NP_001310191.1 | |||
NM_001323263.1 | 216 | Intron | NP_001310192.1 | |||
NM_001323264.1 | 216 | Intron | NP_001310193.1 | |||
NM_052904.3 | 216 | Intron | NP_443136.2 | |||
XM_005266813.4 | 216 | Intron | XP_005266870.1 | |||
XM_017010228.1 | 216 | Intron | XP_016865717.1 | |||
XM_017010229.1 | 216 | Intron | XP_016865718.1 |
NDUFAF4 - NADH:ubiquinone oxidoreductase complex assembly factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014165.3 | 216 | Intron | NP_054884.1 |