Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGCTGTGTCCAGTGTCATCTTC[A/G]TGGTTGTGCCTCTCAGCCTCATCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR2H1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2H1 - olfactory receptor family 2 subfamily H member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318014.1 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | NP_001304943.1 | |
NM_001318022.1 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | NP_001304951.1 | |
NM_030883.4 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | NP_112145.1 | |
XM_011514470.2 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_011512772.1 | |
XM_011514472.2 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_011512774.1 | |
XM_011514473.2 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_011512775.1 | |
XM_011514479.2 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_011512781.1 | |
XM_011514483.2 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_011512785.1 | |
XM_011514484.2 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_011512786.1 | |
XM_017010737.1 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_016866226.1 | |
XM_017010738.1 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_016866227.1 | |
XM_017010739.1 | 2355 | Missense Mutation | ATG,GTG | M,V 205 | XP_016866228.1 | |
XM_017010740.1 | 2355 | Missense Mutation | ATG,GTG | M,V 127 | XP_016866229.1 |
OR5V1 - olfactory receptor family 5 subfamily V member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |