Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTATCGGTTCCCTTAATGGGG[A/G]AAGCTTGTGTGGAAGCTTCTGTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615358 | ||||||||||||||||||||
Literature Links: |
AK9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK9 - adenylate kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145128.2 | 6301 | Missense Mutation | CCC,TCC | P,S 1787 | NP_001138600.2 | |
NM_145025.4 | 6301 | Intron | NP_659462.1 | |||
XM_006715376.3 | 6301 | Missense Mutation | CCC,TCC | P,S 1859 | XP_006715439.2 | |
XM_011535550.2 | 6301 | Missense Mutation | CCC,TCC | P,S 1864 | XP_011533852.1 | |
XM_011535552.2 | 6301 | Missense Mutation | CCC,TCC | P,S 1792 | XP_011533854.1 | |
XM_011535553.2 | 6301 | Missense Mutation | CCC,TCC | P,S 1792 | XP_011533855.1 | |
XM_011535554.2 | 6301 | Missense Mutation | CCC,TCC | P,S 1757 | XP_011533856.1 | |
XM_011535555.2 | 6301 | Intron | XP_011533857.1 | |||
XM_011535556.2 | 6301 | Intron | XP_011533858.1 | |||
XM_011535558.1 | 6301 | Missense Mutation | CCC,TCC | P,S 1068 | XP_011533860.1 | |
XM_011535559.2 | 6301 | Intron | XP_011533861.1 | |||
XM_017010384.1 | 6301 | Intron | XP_016865873.1 | |||
XM_017010385.1 | 6301 | Missense Mutation | CCC,TCC | P,S 1577 | XP_016865874.1 | |
XM_017010386.1 | 6301 | Intron | XP_016865875.1 | |||
XM_017010387.1 | 6301 | Missense Mutation | CCC,TCC | P,S 1410 | XP_016865876.1 | |
XM_017010388.1 | 6301 | Intron | XP_016865877.1 | |||
XM_017010389.1 | 6301 | Missense Mutation | CCC,TCC | P,S 1209 | XP_016865878.1 |