Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGGACGCGCCCCTTCTTGGAACA[A/G]TGTCCCACCACGGAGGAGCTCCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606286 | ||||||||||||||||||||
Literature Links: |
DDX43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX43 - DEAD-box helicase 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018665.2 | 345 | Missense Mutation | ATG,GTG | M,V 1 | NP_061135.2 | |
XM_011535926.1 | 345 | Missense Mutation | ATG,GTG | M,V 1 | XP_011534228.1 | |
XM_011535927.2 | 345 | Intron | XP_011534229.1 | |||
XM_011535928.2 | 345 | UTR 5 | XP_011534230.1 | |||
XM_017011003.1 | 345 | UTR 5 | XP_016866492.1 |
OOEP-AS1 - OOEP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |