Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGATACACTTATTTCCTTTTAT[G/T]CGAACACAAGTATATGAGGGGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYCT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYCT1 - myc target 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025107.2 | 11 | Missense Mutation | ATG,ATT | M,I 1 | NP_079383.2 | |
XM_011536149.2 | 11 | Missense Mutation | ATG,ATT | M,I 1 | XP_011534451.1 | |
XM_011536150.2 | 11 | Missense Mutation | ATG,ATT | M,I 1 | XP_011534452.1 | |
XM_011536151.2 | 11 | Missense Mutation | ATG,ATT | M,I 1 | XP_011534453.1 | |
XM_011536152.2 | 11 | Missense Mutation | ATG,ATT | M,I 1 | XP_011534454.1 | |
XM_017011319.1 | 11 | Missense Mutation | ATG,ATT | M,I 1 | XP_016866808.1 |