Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGTCTGGCTGGCTTCCTGGCTC[A/T]TGACTCCAATAATTTCGAATAGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605732 | ||||||||||||||||||||
Literature Links: |
TNFRSF21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFRSF21 - TNF receptor superfamily member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014452.4 | 2340 | Missense Mutation | AAG,ATG | K,M 635 | NP_055267.1 | |
XM_017010744.1 | 2340 | Intron | XP_016866233.1 |