Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTCATCTTTACTGCTGCTGCT[C/G]TTGGTGGCAAGTGGAGATGCTGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600408 | ||||||||||||||||||||
Literature Links: |
DDR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDR1 - discoidin domain receptor tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202521.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_001189450.1 | |
NM_001202522.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_001189451.1 | |
NM_001202523.1 | 298 | Silent Mutation | CTC,CTG | L,L 31 | NP_001189452.1 | |
NM_001297652.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_001284581.1 | |
NM_001297653.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_001284582.1 | |
NM_001297654.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_001284583.1 | |
NM_001954.4 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_001945.3 | |
NM_013993.2 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_054699.2 | |
NM_013994.2 | 298 | Silent Mutation | CTC,CTG | L,L 13 | NP_054700.2 | |
XM_006715185.2 | 298 | Silent Mutation | CTC,CTG | L,L 31 | XP_006715248.1 | |
XM_011514882.2 | 298 | Silent Mutation | CTC,CTG | L,L 31 | XP_011513184.1 | |
XM_011514883.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_011513185.1 | |
XM_011514884.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_011513186.1 | |
XM_011514885.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_011513187.1 | |
XM_011514886.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_011513188.1 | |
XM_011514887.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_011513189.1 | |
XM_011514888.2 | 298 | Silent Mutation | CTC,CTG | L,L 31 | XP_011513190.1 | |
XM_017011268.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_016866757.1 | |
XM_017011269.1 | 298 | Silent Mutation | CTC,CTG | L,L 13 | XP_016866758.1 |
MIR4640 - microRNA 4640 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |