Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGTTTGTCCTCATGAGCTCCA[A/C]GTCGGCGTGCACCATCTCCCTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602884 | ||||||||||||||||||||
Literature Links: |
GMDS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMDS - GDP-mannose 4,6-dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253846.1 | 1012 | Missense Mutation | NP_001240775.1 | |||
NM_001500.3 | 1012 | Missense Mutation | NP_001491.1 | |||
XM_006715066.2 | 1012 | Intron | XP_006715129.1 | |||
XM_011514500.1 | 1012 | Missense Mutation | XP_011512802.1 | |||
XM_011514502.2 | 1012 | Intron | XP_011512804.1 | |||
XM_011514503.2 | 1012 | Intron | XP_011512805.1 | |||
XM_011514505.1 | 1012 | Intron | XP_011512807.1 | |||
XM_011514506.1 | 1012 | Intron | XP_011512808.1 | |||
XM_011514507.1 | 1012 | Intron | XP_011512809.1 | |||
XM_017010752.1 | 1012 | Missense Mutation | XP_016866241.1 |