Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCTGGGTAGATCCGGGTGCCA[C/T]TGTCTGTTGCCAGGGCCGAAGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC14 - zinc finger DHHC-type containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024630.2 | Intron | NP_078906.2 | ||||
NM_153746.1 | Intron | NP_714968.1 | ||||
XM_005267146.3 | Intron | XP_005267203.1 | ||||
XM_011536120.2 | Intron | XP_011534422.1 | ||||
XM_011536123.2 | Intron | XP_011534425.1 | ||||
XM_017011307.1 | Intron | XP_016866796.1 | ||||
XM_017011308.1 | Intron | XP_016866797.1 | ||||
XM_017011309.1 | Intron | XP_016866798.1 | ||||
XM_017011310.1 | Intron | XP_016866799.1 | ||||
XM_017011311.1 | Intron | XP_016866800.1 | ||||
XM_017011312.1 | Intron | XP_016866801.1 | ||||
XM_017011313.1 | Intron | XP_016866802.1 |