Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCACAGGACTAGGCTCTAAG[C/T]CCCTCATTTCAATGAGGGGCCATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613590 MIM: 613595 | ||||||||||||||||||||
Literature Links: |
BTN2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTN2A1 - butyrophilin subfamily 2 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197233.2 | 602 | Missense Mutation | CCC,TCC | P,S 88 | NP_001184162.1 | |
NM_001197234.2 | 602 | Missense Mutation | CCC,TCC | P,S 149 | NP_001184163.1 | |
NM_007049.4 | 602 | Missense Mutation | CCC,TCC | P,S 149 | NP_008980.1 | |
NM_078476.3 | 602 | Missense Mutation | CCC,TCC | P,S 149 | NP_510961.1 |
BTN3A3 - butyrophilin subfamily 3 member A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC285819 - uncharacterized LOC285819 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |