Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATACTCATGGACAAGTTGGCCC[C/T]CTTGGGAAGGCCATATGAATCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN16 - zinc finger and SCAN domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320555.1 | 590 | Missense Mutation | CCC,CTC | P,L 146 | NP_001307484.1 | |
NM_001320556.1 | 590 | Missense Mutation | CCC,CTC | P,L 117 | NP_001307485.1 | |
NM_001320557.1 | 590 | Missense Mutation | CCC,CTC | P,L 146 | NP_001307486.1 | |
NM_001320558.1 | 590 | Missense Mutation | CCC,CTC | P,L 146 | NP_001307487.1 | |
NM_025231.2 | 590 | Missense Mutation | CCC,CTC | P,L 146 | NP_079507.1 | |
XM_017011324.1 | 590 | Missense Mutation | CCC,CTC | P,L 146 | XP_016866813.1 |
ZSCAN16-AS1 - ZSCAN16 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |