Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGGCGGGAGGCTGTGCGCAAAT[G/T]GTTCCCGGGGGGCCGCCCGGGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611106 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H12D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H12D - zinc finger CCCH-type containing 12D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207360.2 | 1244 | Missense Mutation | CAA,CCA | Q,P 325 | NP_997243.2 |