Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGGTTGATGGATTCTGTGAGGC[A/G]ATTCATGCCGATTGACTGCGCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCM3 - minichromosome maintenance complex component 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270472.1 | 2506 | Missense Mutation | CGC,TGC | R,C 777 | NP_001257401.1 | |
NM_002388.4 | 2506 | Missense Mutation | CGC,TGC | R,C 812 | NP_002379.3 |