Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCACACAATTCTTCAAATTAC[A/G]CAGTGACCTCATAAAAATTCCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606845 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCBLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCBLD1 - discoidin, CUB and LCCL domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173674.2 | 3665 | Intron | NP_775945.1 | |||
XM_005266941.1 | 3665 | Intron | XP_005266998.1 | |||
XM_006715463.1 | 3665 | Intron | XP_006715526.1 | |||
XM_011535771.1 | 3665 | Intron | XP_011534073.1 | |||
XM_011535773.2 | 3665 | Intron | XP_011534075.1 | |||
XM_011535774.2 | 3665 | Intron | XP_011534076.1 | |||
XM_017010775.1 | 3665 | Intron | XP_016866264.1 |
GOPC - golgi associated PDZ and coiled-coil motif containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017408.2 | 3665 | UTR 3 | NP_001017408.1 | |||
NM_020399.3 | 3665 | UTR 3 | NP_065132.1 |