Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGCCCGTATGGATGCGCTGGTG[C/T]CGAATCAAGTGCGTCTTCTTGCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614040 | ||||||||||||||||||||
Literature Links: |
ZNF467 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF467 - zinc finger protein 467 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249959.4 | 980 | Silent Mutation | CGA,CGG | R,R 289 | XP_005250016.1 | |
XM_005249960.4 | 980 | Silent Mutation | CGA,CGG | R,R 289 | XP_005250017.1 | |
XM_005249961.4 | 980 | Silent Mutation | CGA,CGG | R,R 289 | XP_005250018.1 | |
XM_006715864.3 | 980 | Silent Mutation | CGA,CGG | R,R 330 | XP_006715927.1 | |
XM_011515855.2 | 980 | Silent Mutation | CGA,CGG | R,R 540 | XP_011514157.1 | |
XM_011515856.2 | 980 | Silent Mutation | CGA,CGG | R,R 540 | XP_011514158.1 | |
XM_011515857.2 | 980 | Silent Mutation | CGA,CGG | R,R 503 | XP_011514159.1 | |
XM_011515858.1 | 980 | Silent Mutation | CGA,CGG | R,R 289 | XP_011514160.1 | |
XM_017011799.1 | 980 | Silent Mutation | CGA,CGG | R,R 464 | XP_016867288.1 |