Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACTTTCTGCAGTGCCTCTGAGA[A/G]CTTACGCAGATCAGCCGAGTAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 238331 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLD - dihydrolipoamide dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000108.4 | 244 | Missense Mutation | ACT,GCT | T,A 34 | NP_000099.2 | |
NM_001289750.1 | 244 | UTR 5 | NP_001276679.1 | |||
NM_001289751.1 | 244 | Missense Mutation | ACT,GCT | T,A 34 | NP_001276680.1 | |
NM_001289752.1 | 244 | Missense Mutation | ACT,GCT | T,A 34 | NP_001276681.1 |