Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAGGTAGCACTTAGGATATTCT[A/C]GATGGAGCTCAGGGCTTTCTTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611332 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB6 - DnaJ heat shock protein family (Hsp40) member B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005494.2 | Intron | NP_005485.1 | ||||
NM_058246.3 | Intron | NP_490647.1 | ||||
XM_005249515.2 | Intron | XP_005249572.1 | ||||
XM_005249516.2 | Intron | XP_005249573.1 | ||||
XM_006715823.1 | Intron | XP_006715886.1 | ||||
XM_011515704.1 | Intron | XP_011514006.1 | ||||
XM_017011633.1 | Intron | XP_016867122.1 |