Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCCCCGCCCTCCTCTGTCCC[C/T]TCGGCCTCCTTCCGCCGGTGGCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603531 MIM: 615783 MIM: 602186 | ||||||||||||||||||||
Literature Links: |
AP1S1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP1S1 - adaptor related protein complex 1 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4653 - microRNA 4653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAT16 - N-acetyltransferase 16 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VGF - VGF nerve growth factor inducible | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003378.3 | 1517 | Silent Mutation | GAA,GAG | E,E 433 | NP_003369.2 | |
XM_005250561.4 | 1517 | Silent Mutation | GAA,GAG | E,E 433 | XP_005250618.1 | |
XM_011516549.2 | 1517 | Silent Mutation | GAA,GAG | E,E 433 | XP_011514851.1 | |
XM_017012579.1 | 1517 | Silent Mutation | GAA,GAG | E,E 433 | XP_016868068.1 |