Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCGGCGCCCAAGGGCCGCGAAG[C/G]GTTCCGAAGCCAGCGGCGGGAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCYL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHCYL2 - adenosylhomocysteinase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRIP2 - striatin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134336.1 | 141 | Missense Mutation | GCG,GGG | A,G 34 | NP_001127808.1 | |
NM_020704.2 | 141 | Missense Mutation | GCG,GGG | A,G 34 | NP_065755.1 | |
XM_006716069.3 | 141 | Missense Mutation | GCG,GGG | A,G 34 | XP_006716132.1 | |
XM_011516432.2 | 141 | Intron | XP_011514734.1 | |||
XM_011516435.1 | 141 | Missense Mutation | GCG,GGG | A,G 34 | XP_011514737.1 | |
XM_017012470.1 | 141 | Missense Mutation | GCG,GGG | A,G 34 | XP_016867959.1 | |
XM_017012471.1 | 141 | Intron | XP_016867960.1 | |||
XM_017012472.1 | 141 | UTR 5 | XP_016867961.1 |