Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTTCGTGTGACCACTTGGCGCTA[C/T]AGTGAGCAGCAGAGCAGCAGCCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614016 MIM: 602048 | ||||||||||||||||||||
Literature Links: |
DAGLB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAGLB - diacylglycerol lipase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142936.1 | 2017 | Missense Mutation | TAT,TGT | Y,C 487 | NP_001136408.1 | |
NM_139179.3 | 2017 | Missense Mutation | TAT,TGT | Y,C 616 | NP_631918.3 |
RAC1 - ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |