Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGATCTTGTGAATAAATGAGC[A/G]TTCATATCCCTGCAAAGAAAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQUB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IQUB - IQ motif and ubiquitin domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282855.1 | 2792 | Missense Mutation | NP_001269784.1 | |||
NM_001321293.1 | 2792 | Missense Mutation | NP_001308222.1 | |||
NM_178827.4 | 2792 | Missense Mutation | NP_849149.3 | |||
XM_005250161.3 | 2792 | Missense Mutation | XP_005250218.1 | |||
XM_005250162.4 | 2792 | Intron | XP_005250219.1 | |||
XM_011515833.2 | 2792 | Missense Mutation | XP_011514135.1 | |||
XM_011515834.2 | 2792 | Missense Mutation | XP_011514136.1 | |||
XM_017011771.1 | 2792 | Missense Mutation | XP_016867260.1 | |||
XM_017011772.1 | 2792 | Missense Mutation | XP_016867261.1 |