Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTGAGGGTTTGACTGAAGCAC[A/G]GCAGATAAAAATAACCAAATTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC136 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC136 - coiled-coil domain containing 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201372.1 | Intron | NP_001188301.1 | ||||
NM_022742.4 | Intron | NP_073579.4 | ||||
XM_005250534.3 | Intron | XP_005250591.1 | ||||
XM_005250538.1 | Intron | XP_005250595.1 | ||||
XM_011516483.1 | Intron | XP_011514785.1 | ||||
XM_011516484.1 | Intron | XP_011514786.1 | ||||
XM_011516485.1 | Intron | XP_011514787.1 | ||||
XM_011516486.1 | Intron | XP_011514788.1 | ||||
XM_011516487.2 | Intron | XP_011514789.1 | ||||
XM_011516488.2 | Intron | XP_011514790.1 | ||||
XM_011516489.1 | Intron | XP_011514791.1 | ||||
XM_011516490.2 | Intron | XP_011514792.1 | ||||
XM_017012532.1 | Intron | XP_016868021.1 | ||||
XM_017012533.1 | Intron | XP_016868022.1 | ||||
XM_017012534.1 | Intron | XP_016868023.1 | ||||
XM_017012535.1 | Intron | XP_016868024.1 | ||||
XM_017012536.1 | Intron | XP_016868025.1 |