Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCGGCTTGCCCGTTGCTGCTGAC[A/G]TTACGAGACCATTTACTTCAGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610089 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB10 - EF-hand calcium binding domain 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RINT1 - RAD50 interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021930.4 | 1375 | Silent Mutation | ACA,ACG | T,T 654 | NP_068749.3 | |
XM_005250524.3 | 1375 | Silent Mutation | ACA,ACG | T,T 313 | XP_005250581.1 | |
XM_011516458.2 | 1375 | Silent Mutation | ACA,ACG | T,T 313 | XP_011514760.1 | |
XM_017012496.1 | 1375 | Silent Mutation | ACA,ACG | T,T 576 | XP_016867985.1 | |
XM_017012497.1 | 1375 | Silent Mutation | ACA,ACG | T,T 313 | XP_016867986.1 | |
XM_017012498.1 | 1375 | Silent Mutation | ACA,ACG | T,T 313 | XP_016867987.1 |