Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGGGCAGGGGCTATGGCTACCG[C/T]GTGCACCACGACGTTGTCGCCGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC29A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC29A4 - solute carrier family 29 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040661.1 | 1027 | Silent Mutation | CGC,CGT | R,R 282 | NP_001035751.1 | |
NM_001300847.1 | 1027 | Silent Mutation | CGC,CGT | R,R 268 | NP_001287776.1 | |
NM_153247.2 | 1027 | Silent Mutation | CGC,CGT | R,R 282 | NP_694979.2 | |
XM_005249658.4 | 1027 | Silent Mutation | CGC,CGT | R,R 268 | XP_005249715.1 | |
XM_006715667.3 | 1027 | Silent Mutation | CGC,CGT | R,R 282 | XP_006715730.1 | |
XM_006715668.2 | 1027 | Silent Mutation | CGC,CGT | R,R 90 | XP_006715731.1 | |
XM_011515200.2 | 1027 | Silent Mutation | CGC,CGT | R,R 282 | XP_011513502.1 | |
XM_011515201.2 | 1027 | Silent Mutation | CGC,CGT | R,R 282 | XP_011513503.1 |
TNRC18 - trinucleotide repeat containing 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |