Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTTTCAGGGCTCTCTTTACCTA[C/T]GAAGGCAACAGCAATGACATCCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610106 | ||||||||||||||||||||
Literature Links: |
DBNL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBNL - drebrin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014436.2 | 197 | Silent Mutation | TAC,TAT | Y,Y 33 | NP_001014436.1 | |
NM_001122956.1 | 197 | Silent Mutation | TAC,TAT | Y,Y 33 | NP_001116428.1 | |
NM_001284313.1 | 197 | Intron | NP_001271242.1 | |||
NM_001284315.1 | 197 | Missense Mutation | ACG,ATG | T,M 1 | NP_001271244.1 | |
NM_014063.6 | 197 | Silent Mutation | TAC,TAT | Y,Y 33 | NP_054782.2 | |
XM_011515325.1 | 197 | Silent Mutation | TAC,TAT | Y,Y 33 | XP_011513627.1 |
LINC00957 - long intergenic non-protein coding RNA 957 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6837 - microRNA 6837 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASA4CP - RAS p21 protein activator 4C, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |