Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCGGCTCCCTTCTATCATAAT[A/G]CAGAGCCCAGAGCAGAGTAATGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613138 | ||||||||||||||||||||
Literature Links: |
TSPAN12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPAN12 - tetraspanin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012338.3 | 626 | Missense Mutation | CAT,TAT | H,Y 249 | NP_036470.1 | |
XM_005250239.2 | 626 | Missense Mutation | CAT,TAT | H,Y 249 | XP_005250296.1 | |
XM_011515993.1 | 626 | Missense Mutation | CAT,TAT | H,Y 249 | XP_011514295.1 | |
XM_011515994.1 | 626 | Missense Mutation | CAT,TAT | H,Y 249 | XP_011514296.1 | |
XM_017011912.1 | 626 | Missense Mutation | CAT,TAT | H,Y 249 | XP_016867401.1 | |
XM_017011913.1 | 626 | Missense Mutation | CAT,TAT | H,Y 224 | XP_016867402.1 | |
XM_017011914.1 | 626 | Missense Mutation | CAT,TAT | H,Y 122 | XP_016867403.1 |