Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 MIM: 602955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4M1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP4M1 - adaptor related protein complex 4 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004722.3 | 28 | Missense Mutation | GAG,GTG | E,V 31 | NP_004713.2 | |
XM_005250689.4 | 28 | Missense Mutation | GAG,GTG | E,V 31 | XP_005250746.1 | |
XM_005250690.4 | 28 | Missense Mutation | GAG,GTG | E,V 31 | XP_005250747.1 | |
XM_006716175.3 | 28 | Missense Mutation | GAG,GTG | E,V 31 | XP_006716238.1 | |
XM_011516685.2 | 28 | Missense Mutation | GAG,GTG | E,V 31 | XP_011514987.1 | |
XM_017012790.1 | 28 | Intron | XP_016868279.1 | |||
XM_017012791.1 | 28 | UTR 5 | XP_016868280.1 |
COPS6 - COP9 signalosome subunit 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM7 - minichromosome maintenance complex component 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278595.1 | 28 | UTR 5 | NP_001265524.1 | |||
NM_005916.4 | 28 | UTR 5 | NP_005907.3 | |||
NM_182776.2 | 28 | Intron | NP_877577.1 | |||
XM_005250348.3 | 28 | Intron | XP_005250405.1 | |||
XM_017012217.1 | 28 | Intron | XP_016867706.1 |
MIR106B - microRNA 106b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR25 - microRNA 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR93 - microRNA 93 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF6 - TATA-box binding protein associated factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |