Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCAGGGCGGGAGCTGTGCTCA[C/G]ACGGTGAGGGAGATGAAGCTGTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176911 | ||||||||||||||||||||
Literature Links: |
PRKAR1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRKAR1B - protein kinase cAMP-dependent type I regulatory subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164758.1 | 1187 | Silent Mutation | GTC,GTG | V,V 381 | NP_001158230.1 | |
NM_001164759.1 | 1187 | Silent Mutation | GTC,GTG | V,V 381 | NP_001158231.1 | |
NM_001164760.1 | 1187 | Silent Mutation | GTC,GTG | V,V 381 | NP_001158232.1 | |
NM_001164761.1 | 1187 | Silent Mutation | GTC,GTG | V,V 381 | NP_001158233.1 | |
NM_001164762.1 | 1187 | Silent Mutation | GTC,GTG | V,V 381 | NP_001158234.1 | |
NM_002735.2 | 1187 | Silent Mutation | GTC,GTG | V,V 381 | NP_002726.1 | |
XM_011515446.2 | 1187 | Silent Mutation | GTC,GTG | V,V 326 | XP_011513748.1 | |
XM_017012433.1 | 1187 | Silent Mutation | GTC,GTG | V,V 251 | XP_016867922.1 | |
XM_017012434.1 | 1187 | Silent Mutation | GTC,GTG | V,V 229 | XP_016867923.1 |