Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTTCTCCTAAGGTATCTGTATA[A/G]CCAAAGACATGAAGAACACTAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176842 | ||||||||||||||||||||
Literature Links: |
C7orf25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C7orf25 - chromosome 7 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099858.1 | 476 | Silent Mutation | GGC,GGT | G,G 151 | NP_001093328.1 | |
NM_024054.2 | 476 | Silent Mutation | GGC,GGT | G,G 93 | NP_076959.2 | |
XM_005249844.3 | 476 | Silent Mutation | GGC,GGT | G,G 93 | XP_005249901.1 | |
XM_017012596.1 | 476 | Silent Mutation | GGC,GGT | G,G 107 | XP_016868085.1 | |
XM_017012597.1 | 476 | Silent Mutation | GGC,GGT | G,G 93 | XP_016868086.1 |
PSMA2 - proteasome subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |