Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGCCCGCTGCATGGTTGCGCTG[A/G]TGCTTGCGGAGGTGGTCCTTGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613914 | ||||||||||||||||||||
Literature Links: |
ZNF746 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF746 - zinc finger protein 746 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163474.1 | 2153 | Silent Mutation | CAC,CAT | H,H 585 | NP_001156946.1 | |
NM_152557.4 | 2153 | Silent Mutation | CAC,CAT | H,H 584 | NP_689770.3 | |
XM_005249955.2 | 2153 | Silent Mutation | CAC,CAT | H,H 600 | XP_005250012.1 | |
XM_005249956.2 | 2153 | Silent Mutation | CAC,CAT | H,H 599 | XP_005250013.1 | |
XM_005249957.1 | 2153 | Silent Mutation | CAC,CAT | H,H 587 | XP_005250014.1 | |
XM_011515838.1 | 2153 | Silent Mutation | CAC,CAT | H,H 572 | XP_011514140.1 |