Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTGCTTCTCAGAGCTTTACCT[A/G]TTCCACGAGCAAAGCATGTGAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603004 MIM: 611980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBAS - glioblastoma amplified sequence | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS17 - mitochondrial ribosomal protein S17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015969.2 | Intron | NP_057053.1 |