Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGGAGGAGGGACTGCTCTGCC[A/G]GGTGGTTCATTCTCCAGAATTCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM71F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM71F1 - family with sequence similarity 71 member F1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282788.1 | 325 | Missense Mutation | CAG,CGG | Q,R 61 | NP_001269717.1 | |
NM_001282789.1 | 325 | Intron | NP_001269718.1 | |||
NM_032599.3 | 325 | Missense Mutation | CAG,CGG | Q,R 61 | NP_115988.1 | |
XM_017012743.1 | 325 | Missense Mutation | CAG,CGG | Q,R 61 | XP_016868232.1 |