Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGTACTTTGGGTCATGATGA[A/G]GAACATAGCTTGGAATCATCACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605340 | ||||||||||||||||||||
Literature Links: |
CYP3A7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP3A7 - cytochrome P450 family 3 subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000765.4 | 1309 | Missense Mutation | CCT,CTT | P,L 401 | NP_000756.3 |
CYP3A7-CYP3A51P - CYP3A7-CYP3A51P readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256497.2 | 1309 | Missense Mutation | CCT,CTT | P,L 401 | NP_001243426.2 |
ZSCAN25 - zinc finger and SCAN domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145115.2 | 1309 | Intron | NP_660090.2 | |||
XM_005250194.2 | 1309 | Intron | XP_005250251.1 | |||
XM_011515905.2 | 1309 | Intron | XP_011514207.1 | |||
XM_011515906.2 | 1309 | Intron | XP_011514208.1 | |||
XM_011515907.2 | 1309 | Intron | XP_011514209.1 | |||
XM_011515908.2 | 1309 | Intron | XP_011514210.1 | |||
XM_011515909.2 | 1309 | Intron | XP_011514211.1 | |||
XM_011515910.2 | 1309 | Intron | XP_011514212.1 | |||
XM_017011824.1 | 1309 | Intron | XP_016867313.1 | |||
XM_017011825.1 | 1309 | Intron | XP_016867314.1 | |||
XM_017011826.1 | 1309 | Intron | XP_016867315.1 | |||
XM_017011827.1 | 1309 | Intron | XP_016867316.1 |