Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCTTCAACTAACAGGAAGGAAG[G/T]CTTTGGATTGTCTGAGAACCCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611119 | ||||||||||||||||||||
Literature Links: |
KLHL7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL7 - kelch like family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUPL2 - nucleoporin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007342.2 | 1083 | Missense Mutation | GGC,GTC | G,V 94 | NP_031368.1 | |
XM_005249592.1 | 1083 | Missense Mutation | GGC,GTC | G,V 94 | XP_005249649.1 | |
XM_005249593.3 | 1083 | UTR 5 | XP_005249650.1 | |||
XM_011515104.2 | 1083 | Intron | XP_011513406.1 | |||
XM_017011714.1 | 1083 | UTR 5 | XP_016867203.1 | |||
XM_017011715.1 | 1083 | UTR 5 | XP_016867204.1 | |||
XM_017011716.1 | 1083 | UTR 5 | XP_016867205.1 | |||
XM_017011717.1 | 1083 | Intron | XP_016867206.1 | |||
XM_017011718.1 | 1083 | Intron | XP_016867207.1 |