Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCAGGCTTGTGGTCGCGCAGG[C/T]GCACTCCAAATGCTCCTCTAACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173430 | ||||||||||||||||||||
Literature Links: |
PDGFA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDGFA - platelet derived growth factor subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002607.5 | 608 | Missense Mutation | ACC,GCC | T,A 178 | NP_002598.4 | |
NM_033023.4 | 608 | Missense Mutation | ACC,GCC | T,A 178 | NP_148983.1 | |
XM_011515415.1 | 608 | Missense Mutation | ACC,GCC | T,A 223 | XP_011513717.1 | |
XM_011515416.1 | 608 | Missense Mutation | ACC,GCC | T,A 216 | XP_011513718.1 | |
XM_011515417.1 | 608 | Missense Mutation | ACC,GCC | T,A 223 | XP_011513719.1 | |
XM_011515418.1 | 608 | Missense Mutation | ACC,GCC | T,A 216 | XP_011513720.1 | |
XM_011515419.2 | 608 | Missense Mutation | ACC,GCC | T,A 177 | XP_011513721.1 | |
XM_017012289.1 | 608 | Missense Mutation | ACC,GCC | T,A 191 | XP_016867778.1 |