Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACCTGGTTGGATCAAATGCAG[A/G]GGTGTCTCGGGCTCAGGTCCCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ADCK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADCK2 - aarF domain containing kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052853.3 | 1269 | Missense Mutation | GAG,GGG | E,G 289 | NP_443085.2 | |
XM_006716170.3 | 1269 | Missense Mutation | GAG,GGG | E,G 289 | XP_006716233.1 | |
XM_011516675.2 | 1269 | Missense Mutation | GAG,GGG | E,G 289 | XP_011514977.1 |