Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCGTCCAAGATGGCGGCAGAAG[C/G]AGCTGGTGGGAAATACAGAAGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608185 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC4 - exocyst complex component 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037126.1 | 33 | Missense Mutation | GCA,GGA | A,G 5 | NP_001032203.1 | |
NM_021807.3 | 33 | Missense Mutation | GCA,GGA | A,G 5 | NP_068579.3 | |
XM_005250523.4 | 33 | Missense Mutation | GCA,GGA | A,G 5 | XP_005250580.1 | |
XM_017012494.1 | 33 | Intron | XP_016867983.1 | |||
XM_017012495.1 | 33 | Missense Mutation | GCA,GGA | A,G 5 | XP_016867984.1 |