Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCCCTGATGCTGGGTTATCCT[A/G]TAAGTAGCGATCAGGATTAAAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604001 MIM: 601637 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP9 - A-kinase anchoring protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYP51A1 - cytochrome P450 family 51 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000786.3 | Intron | NP_000777.1 | ||||
NM_001146152.1 | Intron | NP_001139624.1 |