Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGTAGATACGAAAATGAGAAGA[A/G]CAATGGAACAGCACACTTTCTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMPCB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PMPCB - peptidase, mitochondrial processing beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004279.2 | 321 | Missense Mutation | AAC,AGC | N,S 96 | NP_004270.2 | |
XM_005250717.3 | 321 | Missense Mutation | AAC,AGC | N,S 96 | XP_005250774.1 | |
XM_006716181.3 | 321 | Missense Mutation | AAC,AGC | N,S 96 | XP_006716244.1 |