Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTTCTCCCATTCCAGCAGCTGG[C/T]TCACCAATCCCCGATTTGGACACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124015 MIM: 616695 MIM: 616550 | ||||||||||||||||||||
Literature Links: |
POR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POR - cytochrome p450 oxidoreductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STYXL1 - serine/threonine/tyrosine interacting-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317785.1 | 951 | Missense Mutation | AAC,AGC | N,S 291 | NP_001304714.1 | |
NM_001317786.1 | 951 | Missense Mutation | AAC,AGC | N,S 291 | NP_001304715.1 | |
NM_001317787.1 | 951 | Missense Mutation | AAC,AGC | N,S 195 | NP_001304716.1 | |
NM_001317788.1 | 951 | UTR 3 | NP_001304717.1 | |||
NM_001317789.1 | 951 | Missense Mutation | AAC,AGC | N,S 153 | NP_001304718.1 | |
NM_016086.2 | 951 | Missense Mutation | AAC,AGC | N,S 291 | NP_057170.1 | |
XM_011516292.2 | 951 | Silent Mutation | GAA,GAG | E,E 253 | XP_011514594.1 | |
XM_011516293.2 | 951 | Missense Mutation | AAC,AGC | N,S 291 | XP_011514595.1 | |
XM_011516300.1 | 951 | Missense Mutation | AAC,AGC | N,S 153 | XP_011514602.1 | |
XM_017012296.1 | 951 | Missense Mutation | AAC,AGC | N,S 329 | XP_016867785.1 | |
XM_017012297.1 | 951 | Silent Mutation | GAA,GAG | E,E 253 | XP_016867786.1 | |
XM_017012298.1 | 951 | Silent Mutation | GAA,GAG | E,E 253 | XP_016867787.1 | |
XM_017012299.1 | 951 | UTR 3 | XP_016867788.1 | |||
XM_017012300.1 | 951 | Missense Mutation | AAC,AGC | N,S 213 | XP_016867789.1 | |
XM_017012301.1 | 951 | Missense Mutation | AAC,AGC | N,S 153 | XP_016867790.1 | |
XM_017012302.1 | 951 | Missense Mutation | AAC,AGC | N,S 153 | XP_016867791.1 |
TMEM120A - transmembrane protein 120A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |