Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTTTATCATGGAAAATCAATTG[A/G]CTAAATCAACTGAAGAACGAACATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606090 MIM: 616403 | ||||||||||||||||||||
Literature Links: |
CROT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CROT - carnitine O-octanoyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143935.1 | 199 | Missense Mutation | ACT,GCT | T,A 6 | NP_001137407.1 | |
NM_001243745.1 | 199 | Missense Mutation | ACT,GCT | T,A 6 | NP_001230674.1 | |
NM_021151.3 | 199 | Missense Mutation | ACT,GCT | T,A 6 | NP_066974.2 | |
XM_011516337.2 | 199 | Missense Mutation | ACT,GCT | T,A 6 | XP_011514639.1 | |
XM_017012370.1 | 199 | UTR 5 | XP_016867859.1 |
TP53TG1 - TP53 target 1 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |