Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGCATTTGCCATCATTGCATC[A/G]TCAACCTGTAAGAATAAGCATATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108370 | ||||||||||||||||||||
Literature Links: |
ASNS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASNS - asparagine synthetase (glutamine-hydrolyzing) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178075.1 | 1358 | Intron | NP_001171546.1 | |||
NM_001178076.1 | 1358 | Intron | NP_001171547.1 | |||
NM_001178077.1 | 1358 | Silent Mutation | GAC,GAT | D,D 411 | NP_001171548.1 | |
NM_001673.4 | 1358 | Intron | NP_001664.3 | |||
NM_133436.3 | 1358 | Silent Mutation | GAC,GAT | D,D 494 | NP_597680.2 | |
NM_183356.3 | 1358 | Intron | NP_899199.2 |